Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol (Spectranalyzed™), Fisher Chemical™

Catalog Number p-216147
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
Safe-Cote™ Amber Glass 
67-56-1
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
32.04
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
887
CHEBI:17790
methanol
CO

For use in the ultraviolet range

TRUSTED_SUSTAINABILITY
Melting Point -98°C
Absorbance Pass Test
Color Colorless
Density 0.791g/cm³
Boiling Point 64.7°C
Quantity 1 L
Flash Point 12°C (53.6°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Assay Percent Range ≥99.8%
Residue after Evaporation 5ppm max.
Titratable Acid 0.0003mEq/g max.
Titratable Base 0.0002mEq/g max.
Linear Formula CH3OH
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55 cP at 20°C
UN Number 1230
Water 0.1% max.
Formula Weight 32.04
Percent Purity ≥99.8%
Grade Also meets ACS specifications
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit