Methanol, ultrapure, Spectrophotometric Grade, >99.8%, Alfa Aesar™

Manufacturer: Alfa Aesar 32435K7

Quantity 4L

 View more versions of this product

Catalog No. AA32435K7

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-56-1
    Physical Form Liquid
    Assay Percent Range >99.8%
    Molecular Formula CH3OH
    Formula Weight 32.04
    MDL Number MFCD00004595
    Chemical Name or Material Methanol
    Grade Spectrophotometric grade

    Specifications

    Formula Weight 32.04
    Quantity 4L
    Color Colorless
    DOT Information DOT Class: 3; Packing Group: II
    Density 0.791g/cm3 at 20°C
    EINECS Number 200-659-6
    Physical Form Liquid
    MDL Number MFCD00004595
    CAS 67-56-1
    Odor Mild
    Vapor Pressure 128 hPa (96mm Hg) at 20°C
    Appearance Colorless liquid
    Assay Percent Range >99.8%
    Molecular Formula CH3OH
    Health Hazard 1 H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.
    Chemical Name or Material Methanol
    Grade Spectrophotometric grade

    H225-H301-H311-H331-H370: Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin. Toxic if inhaled. Causes damage to organs.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit