Methanol, World/GMP, ACS/USP/NF/BP/EP Grade, PHARMCO-AAPER

Manufacturer: Pharmco Products 339WORLDDM55UV

Packaging UV poly drum
Quantity 55 gal.

 View more versions of this product

Catalog No. 16-101-426

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 67-56-1
    Physical Form Liquid
    Molecular Formula CH4O
    Formula Weight 32.04
    Synonym Methyl Alcohol
    Chemical Name or Material Methanol

    Specifications

    Formula Weight 32.04
    Quantity 55 gal.
    Color Clear colorless
    Physical Form Liquid
    Stability Stable under recommended storage conditions.
    CAS 67-56-1
    Synonym Methyl Alcohol
    Description General purpose organic solvent
    Odor Sharp, pungent
    Packaging UV poly drum
    Molecular Formula CH4O
    Chemical Name or Material Methanol

    Order Info

    Minimum Order Quantity: 5

    Minimum Order Quantity: 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit