Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 4-hydroxybenzoate-(ring-13C6), Honeywell Fluka™

Click to view available options
Quantity:
10 mg
1581694-95-2
117065203
1581694-95-2
117065203

Analytical standard, reference material

Ensure you have the information you need to quickly start your analysis, and pass relevant audits:

  • High quality inorganic and organic reference materials produced according to ISO 9001
  • Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
  • Free expert technical support
Physical Form Crystalline
Grade Analytical Standard
Packaging Glass Bottle
Density 1.38 g/cm3
Quantity 10 mg
For Use With (Application) PARABENES