Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methicillin sodium salt, 50mg/mL in distilled water, sterile-filtered

Click to view available options
Quantity:
1mL
5 x 1mL
132-92-3
C17H19N2NaO6S
402.397
MFCD07787409
MGFZNWDWOKASQZ-UMLIZJHQSA-M
23667627
CHEBI:52065
sodium;(2S,5R,6R)-6-[(2,6-dimethoxybenzoyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)C3=C(C=CC=C3OC)OC)C(=O)[O-])C.[Na+]
132-92-3
C17H19N2NaO6S
402.397
MFCD07787409
MGFZNWDWOKASQZ-UMLIZJHQSA-M
23667627
CHEBI:52065
sodium;(2S,5R,6R)-6-[(2,6-dimethoxybenzoyl)amino]-3,3-dimethyl-7-oxo-4-thia-1-azabicyclo[3.2.0]heptane-2-carboxylate
CC1(C(N2C(S1)C(C2=O)NC(=O)C3=C(C=CC=C3OC)OC)C(=O)[O-])C.[Na+]
Physical Form Liquid
Quantity 5 x 1mL
Chemical Name or Material Methicillin sodium salt, 50mg/mL in distilled water, sterile-filtered

RTECSNumber : XH8950000

storageNote1 : -30° to -10°C