Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methionol 99.0+%, TCI America™

Click to view available options
Quantity:
5g
10g
25g
505-10-2
C4H10OS
106.183
MFCD00036560
CZUGFKJYCPYHHV-UHFFFAOYSA-N
methionol,3-methylthiopropanol,3-methylthio-1-propanol,3-hydroxypropyl methyl sulfide,3-methylthio propanol,3-methylmercapto-1-propanol,1-propanol, 3-methylthio,3-methylsulfanyl propan-1-ol,gamma-methylmercaptopropyl alcohol
10448
CHEBI:49019
3-methylsulfanylpropan-1-ol
CSCCCO
505-10-2
C4H10OS
106.183
MFCD00036560
CZUGFKJYCPYHHV-UHFFFAOYSA-N
methionol,3-methylthiopropanol,3-methylthio-1-propanol,3-hydroxypropyl methyl sulfide,3-methylthio propanol,3-methylmercapto-1-propanol,1-propanol, 3-methylthio,3-methylsulfanyl propan-1-ol,gamma-methylmercaptopropyl alcohol
10448
CHEBI:49019
3-methylsulfanylpropan-1-ol
CSCCCO
Boiling Point 136°C
Quantity 10g
Color Yellow
Formula Weight 106.18
Physical Form Clear Liquid at 20°C
Percent Purity ≥99.0% (GC)
Chemical Name or Material Methionol

EINECSNumber : (2)-0469

TSCA : Yes