Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methiothepin maleate, Tocris Bioscience™

Click to view available options
Quantity:
50mg
This item is not returnable. View return policy
19728-88-2
C24H28N2O4S2
472.618
IWDBEHWZGDSFHR-BTJKTKAUSA-N
methiothepin maleate, metitepine maleate, 8-methylthio-10-4-methylpiperazino-10,11-dihydrodibenzo b,f thiepin maleate, 1-10,11-dihydro-8-methylthio dibenzo b,f thiepin-10-yl-4-methylpiperazine maleate, 1-methyl-4-8-methylsulfanyl-10,11-dihydrodibenzo b,f thiepin-10-yl piperazine 2z-but-2-enedioate, prestwick_739, lvp2xbc3ga, unii-lvp2xbc3ga, spectrum1503637, 1-10,11-dihydro-8-methylthio dibenzo z b,f thiepin-10-yl-4-methylpiperazine maleate
5358812
CHEBI:64202
(Z)-but-2-enedioic acid;1-methyl-4-(3-methylsulfanyl-5,6-dihydrobenzo[b][1]benzothiepin-5-yl)piperazine
CN1CCN(CC1)C2CC3=CC=CC=C3SC4=C2C=C(C=C4)SC.C(=CC(=O)O)C(=O)O
This item is not returnable. View return policy
19728-88-2
C24H28N2O4S2
472.618
IWDBEHWZGDSFHR-BTJKTKAUSA-N
methiothepin maleate, metitepine maleate, 8-methylthio-10-4-methylpiperazino-10,11-dihydrodibenzo b,f thiepin maleate, 1-10,11-dihydro-8-methylthio dibenzo b,f thiepin-10-yl-4-methylpiperazine maleate, 1-methyl-4-8-methylsulfanyl-10,11-dihydrodibenzo b,f thiepin-10-yl piperazine 2z-but-2-enedioate, prestwick_739, lvp2xbc3ga, unii-lvp2xbc3ga, spectrum1503637, 1-10,11-dihydro-8-methylthio dibenzo z b,f thiepin-10-yl-4-methylpiperazine maleate
5358812
CHEBI:64202
(Z)-but-2-enedioic acid;1-methyl-4-(3-methylsulfanyl-5,6-dihydrobenzo[b][1]benzothiepin-5-yl)piperazine
CN1CCN(CC1)C2CC3=CC=CC=C3SC4=C2C=C(C=C4)SC.C(=CC(=O)O)C(=O)O

5-HT1 and 5-HT2 antagonist; also has moderate affinity for 5-ht5 and high affinity for 5-HT6 and 5-HT7

5-HT2 antagonist (pKi = 8.7 - 9.2) ; also active as 5-HT1 antagonist (pKi values are 7.1 - 8.5). Also displays affinity for rodent 5-HT5B, 5-HT5A, 5-HT7 and 5-HT6 receptors (pKi values are 6.6, 7.0, 8.4 - 9.0 and 8.7 - 9.4, respectively).
Quantity 50mg
Formula Weight 472.62
Percent Purity >99%
Chemical Name or Material Methiothepin maleate

Recommended Storage : Store at Room Temperature