Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Internal Standard, Acenapthene-d10, SPEX CertiPrep™

Catalog Number 5481is
Click to view available options
Quantity:
1 mL
Packaging:
2mL amber prescored glass ampule
15067-26-2, 67-56-1
C12H10
164.273
CWRYPZZKDGJXCA-WHUVPORUSA-N
177559
1,1,2,2,3,4,5,6,7,8-decadeuterioacenaphthylene
C1CC2=CC=CC3=C2C1=CC=C3
15067-26-2, 67-56-1
C12H10
164.273
CWRYPZZKDGJXCA-WHUVPORUSA-N
177559
1,1,2,2,3,4,5,6,7,8-decadeuterioacenaphthylene
C1CC2=CC=CC3=C2C1=CC=C3

Includes

Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
Acenaphthene-d1015067-26-22000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

CAS Min % 0.2, 99.8
CAS Max % 0.2, 99.8
Packaging 2mL amber prescored glass ampule
Methods For US EPA Method 548.1
Analyte or Component Names Acenaphthene-d10
Solvent or Matrix Methanol
Quantity 1 mL
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
Chemical Name or Material Acenapthene-d10
DOTInformation Nonhazardous
Recommended Storage Ambient

Product intended for laboratory use only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit