Method 552.2 Methyl Esters, SPEX CertiPrep™ Available on GSA/VA Contract for Federal Government customers only.

Manufacturer: SPEX CertiPrepSupplier Diversity Partner 5522MEO

Packaging 2mL amber prescored glass ampule
Quantity 1mL

 View more versions of this product

Catalog No. 5522MEO

  Check Availability
Add to Cart

    Chemical Identifiers

    Chemical Name or Material Methyl Esters Mix

    Specifications

    Quantity 1mL
    For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
    Packaging 2mL amber prescored glass ampule
    Solvent or Matrix MTBE
    Recommended Storage Freezer
    Methods For US EPA Methods 552, 552.2
    Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
    DOT Information Nonhazardous
    Analyte or Component Names 400μg/mL Dalapon methyl ester; 600μg/mL Methyl monochloroacetate; 400μg/mL Methyl mono-bromacetate; 400μg/mL Methyl bromo-chloroacetate; 400μg/mL Methyl bromodi-chlroacetate; 1000μg/mL Methyl chlorodi-bromoacetate; 200μg/mL Methyl dibromoacetate; 1000μg/mL Methyl 2,3-dibromopropionate (surrogate); 600μg/mL Methyl dichloroacetate; 2000μg/mL Methyl tribromoacetate; 200μg/mL Methyl trichloroacetate
    Chemical Name or Material Methyl Esters Mix

    Description

    AnalyteCAS No.Concentration
    Methyl Tribromoacetate3222-05-72000µg/mL
    Methyl Chlorodibromoacetate20428-75-51000µg/mL
    Methyl Chloroacetate96-34-4600µg/mL
    Methyl Dichloroacetate116-54-1600µg/mL
    Dalapon methyl ester17640-02-7400µg/mL
    Methyl Bromoacetate96-32-2400µg/mL
    Methyl Bromochloroacetate20428-74-4400µg/mL
    Methyl Bromodichloroacetate20428-76-6400µg/mL
    Methyl Dibromoacetate6482-26-4200µg/mL
    Methyl Trichloroacetate598-99-2200µg/mL

    Certifications

    Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

    Compliance

    GHS/OSHA Complaint

    Freezer

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit