Learn More
Internal Standard For GC/FID, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Manufacturer: SPEX CertiPrep 8015BI
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
1,1,1,3,3,3-Hexafluoro-2-methyl-2-propanol | 1515-14-6 | 5000μg/mL |
1,1,1,3,3,3-Hexafluoro-2-propanol | 920-66-1 | 5000μg/mL |
2-Chloroacrylonitrile | 920-37-6 | 5000μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. | |
Internal Standard for GC/FID | |
For US EPA Method 8015B | |
H2O |
GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation | |
2mL amber prescored glass ampule | |
2-Chloroacrylonitrile; 1,1,1 | |
1mL |
Safety and Handling
Highly flammable liquid and vapor. May cause cancer. Causes serious eye irritation. May cause drowsiness or dizziness.
DOTInformation : Nonhazardous
Recommended Storage : Ambient
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product intended for laboratory use only