Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyacetic acid, 97+%, ACROS Organics™

Click to view available options
Quantity:
25mL
250mL
1L
Packaging:
Glass bottle
625-45-6
C3H6O3
90.08
MFCD00004308
RMIODHQZRUFFFF-UHFFFAOYSA-N
methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
12251
2-methoxyacetic acid
COCC(=O)O
625-45-6
C3H6O3
90.08
MFCD00004308
RMIODHQZRUFFFF-UHFFFAOYSA-N
methoxyacetic acid, acetic acid, methoxy, methoxyethanoic acid, acetic acid, 2-methoxy, ch3och2cooh, methoxyacetlc acid, unii-f11t1h7q7w, methoxy acetic acid, methoxy-acetic acid, ccris 6518
12251
2-methoxyacetic acid
COCC(=O)O
Flash Point 121°C
Specific Gravity 1.174
Melting Point 7°C
Boiling Point 202°C to 204°C
Refractive Index 1.4153 to 1.4173
Infrared Spectrum Authentic
Color Colorless to Yellow
Quantity 250mL
Density 1.1740g/mL
Packaging Glass bottle
Assay Percent Range 97+%
Linear Formula CH3OCH2CO2H
Beilstein 03, 232
Solubility Information Solubility in water: freely soluble
Formula Weight 90.08
Physical Form Liquid
Percent Purity 97+%
Chemical Name or Material Methoxyacetic acid, 98%
Show More Show Less

GHS H Statement
May cause respiratory irritation.
Causes severe skin burns and eye damage.
Harmful if swallowed.
May damage fertility.
May damage the unborn child.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

EINECSNumber : 210-894-6

RTECSNumber : AI8650000

TSCA : TSCA