Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methoxyacetic acid hydrazide, 95%

Click to view available options
Quantity:
5g
25g
20605-41-8
C3H8N2O2
104.109
MFCD02197887
XEPXDMNZXBUSOI-UHFFFAOYSA-N
methoxyacetic acid hydrazide,methoxyacetohydrazide,acetic acid, methoxy-, hydrazide,2-methoxyethanehydrazide,2-methoxyacetic acid hydrazide,acetic acid,2-methoxy-, hydrazide,methoxymethyl-carboxylic acid hydrazide
2759532
2-methoxyacetohydrazide
COCC(=O)NN
20605-41-8
C3H8N2O2
104.109
MFCD02197887
XEPXDMNZXBUSOI-UHFFFAOYSA-N
methoxyacetic acid hydrazide,methoxyacetohydrazide,acetic acid, methoxy-, hydrazide,2-methoxyethanehydrazide,2-methoxyacetic acid hydrazide,acetic acid,2-methoxy-, hydrazide,methoxymethyl-carboxylic acid hydrazide
2759532
2-methoxyacetohydrazide
COCC(=O)NN
Quantity 25g
Linear Formula CH3OCH2CONHNH2
Beilstein 1903182
Formula Weight 104.11
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Methoxyacetic acid hydrazide