Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyacetone, 96%, ACROS Organics™

Click to view available options
Quantity:
10g
50g
Packaging:
Glass bottle
5878-19-3
C4H8O2
88.11
MFCD00008768
CUZLJOLBIRPEFB-UHFFFAOYSA-N
methoxyacetone,methoxy-2-propanone,2-propanone, 1-methoxy,1-methoxy-2-propanone,1-methoxyacetone,methoxyaceton,methoxymethyl methyl ketone,methoxy acetone,ch3coch2och3
22172
1-methoxypropan-2-one
CC(=O)COC
5878-19-3
C4H8O2
88.11
MFCD00008768
CUZLJOLBIRPEFB-UHFFFAOYSA-N
methoxyacetone,methoxy-2-propanone,2-propanone, 1-methoxy,1-methoxy-2-propanone,1-methoxyacetone,methoxyaceton,methoxymethyl methyl ketone,methoxy acetone,ch3coch2och3
22172
1-methoxypropan-2-one
CC(=O)COC
Flash Point 25°C
Packaging Glass bottle
Boiling Point 118.0°C
Quantity 10g
Specific Gravity 0.95
Density 0.9500g/mL
Color Yellow
Infrared Spectrum Authentic
Refractive Index 1.3970 to 1.3990
Assay Percent Range 95% min. (GC)
Linear Formula CH3OCH2COCH3
Beilstein 01, 822
Solubility Information Solubility in water: miscible
Formula Weight 88.11
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Methoxyacetone
Show More Show Less

GHS H Statement:
Flammable liquid and vapor.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 227-549-