Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Methoxymethyl)diphenylphosphine oxide, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
4455-77-0
C14H15O2P
246.24
MFCD00015451
OEPKDBQOTLDTNC-UHFFFAOYSA-N
methoxymethyl diphenylphosphine oxide,methoxymethyldiphenylphosphine oxide,diphenyl methoxymethyl phosphine oxide,phosphine oxide, methoxymethyl diphenyl,methoxymethyl diphenyl phosphine oxide,phosphine oxide, diphenyl methoxymethyl,methoxymethyl phenyl phosphoroso benzene,acmc-1ahy3,cbmicro_008972
78203
[methoxymethyl(phenyl)phosphoryl]benzene
COCP(=O)(C1=CC=CC=C1)C2=CC=CC=C2
4455-77-0
C14H15O2P
246.24
MFCD00015451
OEPKDBQOTLDTNC-UHFFFAOYSA-N
methoxymethyl diphenylphosphine oxide,methoxymethyldiphenylphosphine oxide,diphenyl methoxymethyl phosphine oxide,phosphine oxide, methoxymethyl diphenyl,methoxymethyl diphenyl phosphine oxide,phosphine oxide, diphenyl methoxymethyl,methoxymethyl phenyl phosphoroso benzene,acmc-1ahy3,cbmicro_008972
78203
[methoxymethyl(phenyl)phosphoryl]benzene
COCP(=O)(C1=CC=CC=C1)C2=CC=CC=C2
For C-C Bond Formation (Olefination)
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color White to Yellow
Melting Point 114.0°C to 117.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Formula Weight 246.24
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material (Methoxymethyl)diphenylphosphine oxide
Show More Show Less

GHS H Statement:
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 224-704-4