Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(Methoxymethyl)triphenylphosphonium chloride, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
500g
Packaging:
Glass bottle
4009-98-7
C20H20ClOP
342.79
MFCD00011800
SJFNDMHZXCUXSA-UHFFFAOYSA-M
methoxymethyl triphenylphosphonium chloride,methoxymethyl triphenylphosphanium chloride,phosphonium, methoxymethyl triphenyl-, chloride,methoxymethyltriphenylphosphonium chloride,methoxymethyl-triphenylphosphonium chloride,methoxymethyl-triphenyl-phosphonium chloride,triphenyl methoxymethyl phosphonium chloride,methoxy methyl triphenyl phosphonium chloride
2723798
methoxymethyl(triphenyl)phosphanium;chloride
COC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
4009-98-7
C20H20ClOP
342.79
MFCD00011800
SJFNDMHZXCUXSA-UHFFFAOYSA-M
methoxymethyl triphenylphosphonium chloride,methoxymethyl triphenylphosphanium chloride,phosphonium, methoxymethyl triphenyl-, chloride,methoxymethyltriphenylphosphonium chloride,methoxymethyl-triphenylphosphonium chloride,methoxymethyl-triphenyl-phosphonium chloride,triphenyl methoxymethyl phosphonium chloride,methoxy methyl triphenyl phosphonium chloride
2723798
methoxymethyl(triphenyl)phosphanium;chloride
COC[P+](C1=CC=CC=C1)(C2=CC=CC=C2)C3=CC=CC=C3.[Cl-]
For C-C Bond Formation (Olefination)
Quantity 25g
Infrared Spectrum Authentic
Boiling Point 413.0°C
Packaging Glass bottle
Melting Point 195.0°C to 197.0°C
Color White
Flash Point >250°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (HPLC)
Linear Formula CH3OCH2P(C6H5)3Cl
Solubility Information Solubility in water: decomposes
Formula Weight 342.79
Physical Form Crystalline Powder
Percent Purity 98%
Chemical Name or Material (Methoxymethyl)triphenylphosphonium chloride, 98%
Show More Show Less

GHS H Statement
Toxic to aquatic life with long lasting effects.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
Avoid release to the environment.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/f

GHS Signal Word: Warning

EINECSNumber : 223-664-5

TSCA : TSCA