Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Methoxypolyethylene Glycol Antibody (HL1020), Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP313681
This item is not returnable.
View return policy
Description
Methoxypolyethylene Glycol Monoclonal antibody specifically detects Methoxypolyethylene Glycol in All species samples. It is validated for Western Blot, ELISASpecifications
| Methoxypolyethylene Glycol | |
| Monoclonal | |
| Unconjugated | |
| Methoxy Polyethylene Glycol, Methoxypoly(ethylene glycol), mPEG | |
| The immunogen used to generate this antibody corresponds to Polyethylene Glycol (methoxy group). | |
| 100 μg | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| HL1020 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| All species | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction