Learn More
Sigma Aldrich Methoxypolyethylene Glycol Propyl Amine 1 000
Shop All MilliporeSigma Sigma Organic Chemistry Products
Description
Specifications
Specifications
| Linear Formula | CH3(OCH2CH2)nOCH2CH2CH2NH2 |
| Quantity | 1 g |
| Synonym | PEG amine; Poly(ethylene) glycol amine; mPEG NH2; mPEG amine |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.