Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyresorufin, Tocris Bioscience™

Click to view available options
Quantity:
10mg
This item is not returnable. View return policy
5725-89-3
C13H9NO3
227.219
KNYYMGDYROYBRE-UHFFFAOYSA-N
7-methoxy-3h-phenoxazin-3-one,methoxyresorufin,resorufin methyl ether,7-methoxyresorufin,o7-methylresorufin,o-methylresorufin,3h-phenoxazin-3-one, 7-methoxy,7-methoxy resorufin,resorufin 7-o-methyl ether,us9216974, methoxy-resorufin
119220
7-methoxyphenoxazin-3-one
COC1=CC2=C(C=C1)N=C3C=CC(=O)C=C3O2
This item is not returnable. View return policy
5725-89-3
C13H9NO3
227.219
KNYYMGDYROYBRE-UHFFFAOYSA-N
7-methoxy-3h-phenoxazin-3-one,methoxyresorufin,resorufin methyl ether,7-methoxyresorufin,o7-methylresorufin,o-methylresorufin,3h-phenoxazin-3-one, 7-methoxy,7-methoxy resorufin,resorufin 7-o-methyl ether,us9216974, methoxy-resorufin
119220
7-methoxyphenoxazin-3-one
COC1=CC2=C(C=C1)N=C3C=CC(=O)C=C3O2

Fluorometric CYP450 substrate

Fluorometric substrate of cytochrome P450; displays preferential metabolism by rodent cytochrome P4501A2 (CYP1A2).
Quantity 10mg
Formula Weight 227.22
Percent Purity >98%
Chemical Name or Material Methoxyresorufin

Recommended Storage : Store at -20°C