Alfa Aesar™ Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate, 98%

Manufacturer: Alfa Aesar™ H31888-MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH31888MD

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 7355-18-2
    Molecular Formula C15H20O11
    Formula Weight 376.32g/mol
    MDL Number MFCD00069834
    Synonym MTAG
    Chemical Name or Material Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate

    Specifications

    Melting Point 177° to 181°C
    Quantity 250mg
    Odor Odorless
    Optical Rotation +8.5° (c=1 in Chloroform)
    CAS 7355-18-2
    Molecular Formula C15H20O11
    Formula Weight 376.32g/mol
    MDL Number MFCD00069834
    Synonym MTAG
    Chemical Name or Material Methyl 1,2,3,4-tetra-O-acetyl-beta-D-glucuronate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit