Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hydrochloride, 97%, Maybridge

Click to view available options
Quantity:
1g
Packaging:
Amber Glass Bottle
78348-26-2
C11H14ClNO2
227.688
MFCD02180877
GZCFAFBVBOGXRN-UHFFFAOYSA-N
methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hydrochloride,methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hcl,1,2,3,4-tetrahydro-quinoline-2-carboxylic acid methyl ester hcl,methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate, chloride,2-methoxycarbonyl-1,2,3,4-tetrahydroquinoline hydrochloride,methyl 1,2,3,4-tetrahydroquinolin-2-carboxylate hydrochloride,1,2,3,4-tetrahydroquinoline-2-carboxylic acid methyl ester hydrochloride
2747398
methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate;hydrochloride
COC(=O)C1CCC2=CC=CC=C2N1.Cl
78348-26-2
C11H14ClNO2
227.688
MFCD02180877
GZCFAFBVBOGXRN-UHFFFAOYSA-N
methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hydrochloride,methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hcl,1,2,3,4-tetrahydro-quinoline-2-carboxylic acid methyl ester hcl,methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate, chloride,2-methoxycarbonyl-1,2,3,4-tetrahydroquinoline hydrochloride,methyl 1,2,3,4-tetrahydroquinolin-2-carboxylate hydrochloride,1,2,3,4-tetrahydroquinoline-2-carboxylic acid methyl ester hydrochloride
2747398
methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate;hydrochloride
COC(=O)C1CCC2=CC=CC=C2N1.Cl
Packaging Amber Glass Bottle
Quantity 1g
CAS Min % 97.0
Assay Percent Range 97%
Formula Weight 227.69
Percent Purity 97%
Chemical Name or Material Methyl 1,2,3,4-tetrahydroquinoline-2-carboxylate hydrochloride

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning