Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 1-aminocyclobutanecarboxylate hydrochloride, 95%

Click to view available options
Quantity:
250mg
1g
5g
215597-35-6
C6H11NO2
129.159
MFCD04115288
SGXOPTPGZBLQAY-UHFFFAOYSA-N
methyl 1-aminocyclobutanecarboxylate,1-amino-cyclobutanecarboxylic acid methyl ester,1-aminocyclobutanecarboxylic acid methyl ester,cyclobutanecarboxylic acid, 1-amino-, methyl ester,1-amino-cyclobutanecarboxylicacidmethylester,methyl 1-amino-cyclobutane-1-carboxylate,1-aminocyclobutane-1-carboxylic acid methyl ester,cyclobutanecarboxylicacid, 1-amino-, methyl ester
1502033
methyl 1-aminocyclobutane-1-carboxylate
COC(=O)C1(CCC1)N
215597-35-6
C6H11NO2
129.159
MFCD04115288
SGXOPTPGZBLQAY-UHFFFAOYSA-N
methyl 1-aminocyclobutanecarboxylate,1-amino-cyclobutanecarboxylic acid methyl ester,1-aminocyclobutanecarboxylic acid methyl ester,cyclobutanecarboxylic acid, 1-amino-, methyl ester,1-amino-cyclobutanecarboxylicacidmethylester,methyl 1-amino-cyclobutane-1-carboxylate,1-aminocyclobutane-1-carboxylic acid methyl ester,cyclobutanecarboxylicacid, 1-amino-, methyl ester
1502033
methyl 1-aminocyclobutane-1-carboxylate
COC(=O)C1(CCC1)N
Applications
Used as a primary and secondary intermediate for pharmaceutical and organic synthesis.

Solubility
Slightly soluble in water.

Notes
Air Sensitive. Store away form air. Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
Sensitivity Air Sensitive
Quantity 1g
Beilstein 5615121
Formula Weight 165.62
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Methyl 1-aminocyclobutanecarboxylate hydrochloride

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning