Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 97%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
3939-01-3
C14H17NO3·HCl
283.76
MFCD00012799
BRADBAOVPACOQQ-UHFFFAOYNA-N
methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate, hcl, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5
107479
3939-01-3
C14H17NO3·HCl
283.76
MFCD00012799
BRADBAOVPACOQQ-UHFFFAOYNA-N
methyl 1-benzyl-4-oxopiperidine-3-carboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate hydrochloride, methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 1-benzyl-3-carbomethoxy-4-piperidone hydrochloride, 1-benzyl-3-methoxycarbonyl-4-piperidone hydrochloride, methyl 1-benzyl-4-oxo-3-piperidinecarboxylate, hcl, methyl 1-benzyl-4-oxopiperidine-3-carboxylate hcl, 1-benzyl-3-methoxycarbonyl-4-piperidone hcl, pubchem12956, acmc-1cll5
107479
Quantity 100g
Melting Point 172°C
Infrared Spectrum Authentic
Packaging Glass bottle
Color Pink to White
Assay Percent Range 96% min. (HPLC)
Formula Weight 283.76
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material Methyl 1-benzyl-4-oxo-3-piperidine-carboxylate hydrochloride, 97%

HYGROSCOPIC

EINECSNumber : 223-522-2