Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
Packaging:
Glass bottle
51535-00-3
C13H15NO3
233.27
MFCD00003196
WTRWSSDZHQOPJI-UHFFFAOYSA-N
methyl 1-benzyl-5-oxo-3-pyrrolidinecarboxylate,1-benzyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester,methyl 1-benzyl-5-oxo-pyrrolidine-3-carboxylate,methyl1-benzyl-5-oxo-3-pyrrolidinecarboxylate,3-pyrrolidinecarboxylic acid, 5-oxo-1-phenylmethyl-, methyl ester,methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate,1-benzyl-5-oxo-3-pyrrolidinecarboxylic acid methyl ester,methyl 5-oxo-1-benzylpyrrolidine-3-carboxylate,pubchem17184,methyl-1-benzyl-5-oxopyrrolidin-3-carboxylat
98909
methyl 1-benzyl-5-oxopyrrolidine-3-carboxylate
COC(=O)C1CC(=O)N(C1)CC2=CC=CC=C2
51535-00-3
C13H15NO3
233.27
MFCD00003196
WTRWSSDZHQOPJI-UHFFFAOYSA-N
methyl 1-benzyl-5-oxo-3-pyrrolidinecarboxylate,1-benzyl-5-oxo-pyrrolidine-3-carboxylic acid methyl ester,methyl 1-benzyl-5-oxo-pyrrolidine-3-carboxylate,methyl1-benzyl-5-oxo-3-pyrrolidinecarboxylate,3-pyrrolidinecarboxylic acid, 5-oxo-1-phenylmethyl-, methyl ester,methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate,1-benzyl-5-oxo-3-pyrrolidinecarboxylic acid methyl ester,methyl 5-oxo-1-benzylpyrrolidine-3-carboxylate,pubchem17184,methyl-1-benzyl-5-oxopyrrolidin-3-carboxylat
98909
methyl 1-benzyl-5-oxopyrrolidine-3-carboxylate
COC(=O)C1CC(=O)N(C1)CC2=CC=CC=C2
Infrared Spectrum Authentic
Melting Point 62°C to 66°C
Quantity 5g
Color White to Yellow
Packaging Glass bottle
Assay Percent Range 97.5% min. (GC)
Formula Weight 233.27
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Methyl 1-benzyl-5-oxo-3-pyrrolidine-carboxylate, 98%

EINECSNumber : 257-257-9