Learn More
Alfa Aesar™ Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate
Manufacturer:  Alfa Aesar™ H2783203
Quantity 1g
 View more versions of this product
Catalog No. AAH2783203
  Check Availability
Add to cart
Specifications Description & Specifications
Chemical Identifiers
126937-42-6
378.43
(+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid methyl ester; Piperazine-1,2,4-tricarboxylic acid 1-benzyl 4-tert-butyl 2-methyl triester
C19H26N2O6
MFCD03426296
Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate
Specifications
126937-42-6
378.43
(+/-)-1-Benzyloxycarbonyl-4-Boc-piperazine-2-carboxylic acid methyl ester; Piperazine-1,2,4-tricarboxylic acid 1-benzyl 4-tert-butyl 2-methyl triester
Methyl (+/-)-1-benzyloxycarbonyl-4-Boc-piperazine-2-carboxylate
C19H26N2O6
MFCD03426296
1g
Safety and Handling
SDS & Safety Documents
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning