Learn More
Methyl 1-cyclohexene-1-carboxylate, 97%, Thermo Scientific Chemicals
Description
Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Methyl 1-cyclohexene-1-carboxylate is used in the diastereoselective synthesis of cis-1,2-dialkenylcyclopropanols and methyl -7,7-dimethyl-9-oxo-1,3,4,4a,6,7,8,9,9b-decahydrodibenzo[b,d]furan-4a-carboxylate.
Solubility
Immiscible with water.
Notes
Incompatible with strong oxidizing agents and bases.
Specifications
Density | 1.03 |
Boiling Point | 190°C to 192°C |
Flash Point | 73°C (165°F) |
Refractive Index | 1.477 |
Quantity | 25 g |
Solubility Information | Immiscible with water. |
Formula Weight | 140.18 |
Percent Purity | 97% |
Chemical Name or Material | Methyl 1-cyclohexene-1-carboxylate |
Safety and Handling
Hazard Category | H227 |
Hazard Statement | GHS H Statement H227 Combustible liquid. |
Precautionary Statement | P210-P280-P370+P378q-P501c |
EINECSNumber | 242-331-5 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.