Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-methylcyclopropyl ketone, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
1567-75-5
C6H10O
98.14
OQBCJXUAQQMTRW-UHFFFAOYSA-N
methyl 1-methylcyclopropyl ketone,1-1-methylcyclopropyl ethan-1-one,1-1-methylcyclopropyl ethanone,1-acetyl-1-methylcyclopropane,ethanone, 1-1-methylcyclopropyl,1-methylcyclopropyl methyl ketone,methyl 1-methyl cyclopropyl ketone,1-1-methyl-cyclopropyl-ethanone,methyl1-methylcyclopropylketone
74067
1-(1-methylcyclopropyl)ethanone
CC(=O)C1(CC1)C
1567-75-5
C6H10O
98.14
OQBCJXUAQQMTRW-UHFFFAOYSA-N
methyl 1-methylcyclopropyl ketone,1-1-methylcyclopropyl ethan-1-one,1-1-methylcyclopropyl ethanone,1-acetyl-1-methylcyclopropane,ethanone, 1-1-methylcyclopropyl,1-methylcyclopropyl methyl ketone,methyl 1-methyl cyclopropyl ketone,1-1-methyl-cyclopropyl-ethanone,methyl1-methylcyclopropylketone
74067
1-(1-methylcyclopropyl)ethanone
CC(=O)C1(CC1)C
Refractive Index 1.4330 to 1.4350
Flash Point 24°C
Density 0.8950g/mL
Packaging Glass bottle
Quantity 25g
Boiling Point 125.0°C to 128.0°C
Color Colorless to Yellow
Specific Gravity 0.895
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula CH3C3H4COCH3
Solubility Information Solubility in water: insoluble.
Formula Weight 98.14
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material Methyl 1-methylcyclopropyl ketone
Show More Show Less