Alfa Aesar™ Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate, 96%

Manufacturer: Alfa Aesar™ H2575706

Quantity 5g

 View more versions of this product

Catalog No. AAH2575706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 208242-25-5
    Molecular Formula C6H7F3O2
    Formula Weight 168.12
    MDL Number MFCD06801321
    UN Number UN3272
    Synonym 1-Trifluoromethyl-1-cyclopropanecarboxylic acid methyl ester
    Chemical Name or Material Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate

    Specifications

    Formula Weight 168.12
    Molecular Formula C6H7F3O2
    MDL Number MFCD06801321
    UN Number UN3272
    Synonym 1-Trifluoromethyl-1-cyclopropanecarboxylic acid methyl ester
    Boiling Point 115°C
    Quantity 5g
    Flash Point 42°C (108°F)
    Health Hazard 2 GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: ESTERS, N.O.S.
    CAS 208242-25-5
    Health Hazard 1 Warning
    Chemical Name or Material Methyl 1-trifluoromethyl-1-cyclopropanecarboxylate
    Assay 96%

    GHS H Statement
    H226-H315-H319-H335
    Flammable liquid and vapour.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit