Alfa Aesar™ Methyl 10-undecynoate, 96%

Manufacturer: Alfa Aesar™ H5345314

Quantity 25g

 View more versions of this product

Catalog No. AAH5345314

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2777-66-4
    Molecular Formula C12H20O2
    Linear Formula HC≡C(CH2)8CO2CH3
    Formula Weight 196.29
    MDL Number MFCD00190208
    Synonym 10-Undecynoic acid methyl ester
    Chemical Name or Material Methyl 10-undecynoate

    Specifications

    Formula Weight 196.29
    Molecular Formula C12H20O2
    Linear Formula HC≡C(CH2)8CO2CH3
    MDL Number MFCD00190208
    CAS 2777-66-4
    Synonym 10-Undecynoic acid methyl ester
    Boiling Point 124°C (10mmHg)
    Quantity 25g
    Chemical Name or Material Methyl 10-undecynoate
    Assay 96%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit