Learn More
Alfa Aesar™ Methyl (+/-)-2,2-dimethyl-1,3-dioxolane-4-carboxylate, 97+%
Manufacturer:  Alfa Aesar™ H3378103
Quantity 1g
 View more versions of this product
Catalog No. AAH3378103
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications
Description
Applications
Methyl (+/-)-2,2-dimethyl-1,3-dioxolane-4-carboxylate is used as pharmaceutical intermediate.
Notes
Store in a cool, dry place, in a well sealed container. Store away from oxidizing agents.
Description & Specifications
Chemical Identifiers
108865-84-5
160.17
(+/-)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic acid methyl ester; Methyl 2,3-O-isopropylidene-DL-glycerate
C7H12O4
MFCD00010078
Methyl (+/-)-2,2-dimethyl-1,3-dioxolane-4-carboxylate
Specifications
108865-84-5
160.17
(+/-)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic acid methyl ester; Methyl 2,3-O-isopropylidene-DL-glycerate
1g
Methyl (+/-)-2,2-dimethyl-1,3-dioxolane-4-carboxylate
C7H12O4
MFCD00010078
70° to 75°C (10mmHg)
173°C (343°F)
≥97%