Learn More
Alfa Aesar™ Methyl (+/-)-2,2-dimethyl-1,3-dioxolane-4-carboxylate, 97+%
Manufacturer:  Alfa Aesar™ H33781-09
Quantity 10g
 View more versions of this product
Catalog No. AAH3378109
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Specifications Description & Specifications
Chemical Identifiers
108865-84-5
160.17
(+/-)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic acid methyl ester; Methyl 2,3-O-isopropylidene-DL-glycerate
C7H12O4
MFCD00010078
Methyl (+/-)-2,2-dimethyl-1,3-dioxolane-4-carboxylate
Specifications
108865-84-5
160.17
(+/-)-2,2-Dimethyl-1,3-dioxolane-4-carboxylic acid methyl ester; Methyl 2,3-O-isopropylidene-DL-glycerate
10g
Methyl (+/-)-2,2-dimethyl-1,3-dioxolane-4-carboxylate
C7H12O4
MFCD00010078
70° to 75°C (10mmHg)
173°C (343°F)
≥97%