Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,2-dimethyl-3-hydroxypropionate, 98+%, ACROS Organics™

Click to view available options
Quantity:
25g
100g
Packaging:
Glass bottle
14002-80-3
C6H12O3
132.16
MFCD00009707
KJRFTNVYOAGTHK-UHFFFAOYSA-N
hydroxypivalic acid methyl ester,methyl 3-hydroxypivalate,methyl hydroxypivalate,methyl 2,2-dimethyl-3-hydroxypropionate,hydroxypivalicacidmethylester,propanoic acid, 3-hydroxy-2,2-dimethyl-, methyl ester,2,2-dimethyl-3-hydroxypropionic acid methyl ester,3-hydroxy-2,2-dimethyl-propionic acid methyl ester,methyl 2,2-dimethyl-3-hydroxypropanoate,methyl hydroxylpivalate
84152
methyl 3-hydroxy-2,2-dimethylpropanoate
CC(C)(CO)C(=O)OC
14002-80-3
C6H12O3
132.16
MFCD00009707
KJRFTNVYOAGTHK-UHFFFAOYSA-N
hydroxypivalic acid methyl ester,methyl 3-hydroxypivalate,methyl hydroxypivalate,methyl 2,2-dimethyl-3-hydroxypropionate,hydroxypivalicacidmethylester,propanoic acid, 3-hydroxy-2,2-dimethyl-, methyl ester,2,2-dimethyl-3-hydroxypropionic acid methyl ester,3-hydroxy-2,2-dimethyl-propionic acid methyl ester,methyl 2,2-dimethyl-3-hydroxypropanoate,methyl hydroxylpivalate
84152
methyl 3-hydroxy-2,2-dimethylpropanoate
CC(C)(CO)C(=O)OC
Boiling Point 177°C to 178°C
Flash Point 76°C
Specific Gravity 1.04
Density 1.0400g/mL
Quantity 25g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Colorless
Assay Percent Range 98% min. (GC)
Linear Formula HOCH2C(CH3)2CO2CH3
Beilstein 03, 331
Formula Weight 132.16
Physical Form Liquid
Percent Purity 98+%
Chemical Name or Material Methyl 2,2-dimethyl-3-hydroxypropionate
Show More Show Less

GHS H Statement
Causes serious eye irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
If eye irritation persists: Get medical advice/attention.

GHS Signal Word: Warning

EINECSNumber : 237-805-3