Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2,3,6-tri-O-benzoyl-alpha-D-galactopyranoside, 98%

Click to view available options
Quantity:
1g
5g
3601-36-3
C28H26O9
506.507
MFCD06300842
WXFFEILSURAFKL-SUWSLWCISA-N
methyl 2,3,6-tri-o-benzoyl-alpha-d-galactopyranoside,2s,3r,4s,5s,6r-6-benzoyloxy methyl-5-hydroxy-2-methoxytetrahydro-2h-pyran-3,4-diyl dibenzoate,methyl d-galactopyranoside 2,3,6-tribenzoate,methyl 2,3,6-tri-o-benzoyl-a-d-galactopyranoside,methyl 2,3,6-tri-o-benzoyl-,a-d-galactopyranoside,methyl 2-o,3-o,6-o-tribenzoyl-alpha-d-galactopyranoside,2s,3r,4s,5s,6r-4-benzoyloxy-6-benzoyloxy methyl-5-hydroxy-2-methoxyoxan-3-yl benzoate,2r,3s,4s,5r,6s-4,5-bis benzoyloxy-3-hydroxy-6-methoxyoxan-2-yl methyl benzoate
10929259
[(2R,3S,4S,5R,6S)-4,5-dibenzoyloxy-3-hydroxy-6-methoxyoxan-2-yl]methyl benzoate
COC1C(C(C(C(O1)COC(=O)C2=CC=CC=C2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
3601-36-3
C28H26O9
506.507
MFCD06300842
WXFFEILSURAFKL-SUWSLWCISA-N
methyl 2,3,6-tri-o-benzoyl-alpha-d-galactopyranoside,2s,3r,4s,5s,6r-6-benzoyloxy methyl-5-hydroxy-2-methoxytetrahydro-2h-pyran-3,4-diyl dibenzoate,methyl d-galactopyranoside 2,3,6-tribenzoate,methyl 2,3,6-tri-o-benzoyl-a-d-galactopyranoside,methyl 2,3,6-tri-o-benzoyl-,a-d-galactopyranoside,methyl 2-o,3-o,6-o-tribenzoyl-alpha-d-galactopyranoside,2s,3r,4s,5s,6r-4-benzoyloxy-6-benzoyloxy methyl-5-hydroxy-2-methoxyoxan-3-yl benzoate,2r,3s,4s,5r,6s-4,5-bis benzoyloxy-3-hydroxy-6-methoxyoxan-2-yl methyl benzoate
10929259
[(2R,3S,4S,5R,6S)-4,5-dibenzoyloxy-3-hydroxy-6-methoxyoxan-2-yl]methyl benzoate
COC1C(C(C(C(O1)COC(=O)C2=CC=CC=C2)O)OC(=O)C3=CC=CC=C3)OC(=O)C4=CC=CC=C4
Melting Point 143°C
Color White
Quantity 1g
Optical Rotation +121° (c=1 in Chloroform)
Formula Weight 506.5
Physical Form Powder
Percent Purity 98%
Chemical Name or Material Methyl 2,3,6-tri-O-benzoyl-alpha-D-galactopyranoside

TSCA : No

Recommended Storage : Keep cold