Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,3-dichlorobenzoate, 97%, ACROS Organics™

Click to view available options
Quantity:
25mL
Packaging:
Glass bottle
2905-54-6
C8H6Cl2O2
205.034
MFCD00061099
RFVULUSGHFRDHJ-UHFFFAOYSA-N
2,3-dichlorobenzoic methylester,benzoic acid, 2,3-dichloro-, methyl ester,2,3-dichlorobenzoic acid methyl ester,methyl2,3-dichlorobenzoate,acmc-1cfez,methyl-2,3-dichlorobenzoate
76189
methyl 2,3-dichlorobenzoate
COC(=O)C1=C(C(=CC=C1)Cl)Cl
2905-54-6
C8H6Cl2O2
205.034
MFCD00061099
RFVULUSGHFRDHJ-UHFFFAOYSA-N
2,3-dichlorobenzoic methylester,benzoic acid, 2,3-dichloro-, methyl ester,2,3-dichlorobenzoic acid methyl ester,methyl2,3-dichlorobenzoate,acmc-1cfez,methyl-2,3-dichlorobenzoate
76189
methyl 2,3-dichlorobenzoate
COC(=O)C1=C(C(=CC=C1)Cl)Cl
Density 1.3600g/mL
Packaging Glass bottle
Melting Point 33°C to 38°C
Infrared Spectrum Authentic
Quantity 25mL
Color White
Assay Percent Range 97%
Linear Formula (Cl)2C6H3COOCH3
Formula Weight 205.03
Physical Form Crystals
Percent Purity ≥96.0%
Chemical Name or Material Methyl 2, 3-dichlorobenzoate, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

TSCA : TSCA