Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ Methyl 2,3-dichloropropionate, 98%
Click to view available options
Quantity:
25g
100g
500g
Description
Applications
Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.
Solubility
Soluble in ether, acetone, ethanol.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
Methyl 2,3-dichloropropionate can be used to produce 2-chloro-acrylic acid methyl ester at Heating. It is used as pharmaceutical intermediate.
Solubility
Soluble in ether, acetone, ethanol.
Notes
Store in cool, dry place in tightly closed containers. Store away from strong oxidizing agents, strong bases.
Specifications
Refractive Index | 1.453 |
Density | 1.326 |
Flash Point | 87°C (188°F) |
Boiling Point | 91°C to 92°C (50mmHg) |
Quantity | 500g |
UN Number | UN3265 |
Beilstein | 1701891 |
Solubility Information | Soluble in ether,acetone,ethanol. |
Formula Weight | 157 |
Percent Purity | 98% |
Show More |
Safety and Handling
GHS H Statement
H314-H318-H227
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H227-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 222-940-2
TSCA : Yes
Recommended Storage : Ambient temperatures