Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-dichloropyrimidine-6-carboxylate, 98%, Acros Organics

Click to view available options
Quantity:
5g
25g
6299-85-0
C6H4Cl2N2O2
207.02
DQNQNLWKAGZNIT-UHFFFAOYSA-N
methyl 2,4-dichloropyrimidine-6-carboxylate,methyl 2,4-dichloro-pyrimidine-6-carboxylate,2,6-dichloropyrimidine-4-carboxylic acid methyl ester,4-pyrimidinecarboxylic acid, 2,6-dichloro-, methyl ester,2,6-dichloro-4-pyrimidinecarboxylicacid methyl ester,t6n cnj bg dvo1 fg wln,zlchem 33,pubchem23452,methyl2,6-dichloropyrimidine-4-carboxylate,2,4-dichloro-6-methoxycarbonyl pyrimidine
239689
methyl 2,6-dichloropyrimidine-4-carboxylate
COC(=O)C1=CC(=NC(=N1)Cl)Cl
6299-85-0
C6H4Cl2N2O2
207.02
DQNQNLWKAGZNIT-UHFFFAOYSA-N
methyl 2,4-dichloropyrimidine-6-carboxylate,methyl 2,4-dichloro-pyrimidine-6-carboxylate,2,6-dichloropyrimidine-4-carboxylic acid methyl ester,4-pyrimidinecarboxylic acid, 2,6-dichloro-, methyl ester,2,6-dichloro-4-pyrimidinecarboxylicacid methyl ester,t6n cnj bg dvo1 fg wln,zlchem 33,pubchem23452,methyl2,6-dichloropyrimidine-4-carboxylate,2,4-dichloro-6-methoxycarbonyl pyrimidine
239689
methyl 2,6-dichloropyrimidine-4-carboxylate
COC(=O)C1=CC(=NC(=N1)Cl)Cl
Melting Point 55°C to 56°C
Quantity 25g
Assay Percent Range 97.5% min. (GC)
Formula Weight 207.02
Percent Purity 98%
Chemical Name or Material Methyl 2, 4-dichloropyrimidine-6-carboxylate

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and ke

GHS Signal Word: Warning