Alfa Aesar™ Methyl 2,4-difluorobenzoate, 98%

Manufacturer: Alfa Aesar™ H3216114

Quantity 25g

 View more versions of this product

Catalog No. AAH3216114

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 106614-28-2
    Molecular Formula C8H6F2O2
    Formula Weight 172.13
    MDL Number MFCD03093799
    Synonym 2,4-Difluorobenzoic acid methyl ester
    Chemical Name or Material Methyl 2,4-difluorobenzoate

    Specifications

    Formula Weight 172.13
    Molecular Formula C8H6F2O2
    MDL Number MFCD03093799
    CAS 106614-28-2
    Synonym 2,4-Difluorobenzoic acid methyl ester
    Quantity 25g
    Chemical Name or Material Methyl 2,4-difluorobenzoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit