Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-dihydroxybenzoate, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
2150-47-2
C8H8O4
168.15
MFCD00002276
IIFCLXHRIYTHPV-UHFFFAOYSA-N
2,4-dihydroxybenzoic acid methyl ester,benzoic acid, 2,4-dihydroxy-, methyl ester,methyl beta-resorcylate,methyl-2,4-dihydroxybenzoate,unii-xe68bu3oyj,xe68bu3oyj,.beta.-resorcylic acid, methyl ester,beta-resorcylic acid methyl ester,methyl benzoate, 2,4-dihydroxy,methyl b-resorcylate
16523
methyl 2,4-dihydroxybenzoate
COC(=O)C1=C(C=C(C=C1)O)O
2150-47-2
C8H8O4
168.15
MFCD00002276
IIFCLXHRIYTHPV-UHFFFAOYSA-N
2,4-dihydroxybenzoic acid methyl ester,benzoic acid, 2,4-dihydroxy-, methyl ester,methyl beta-resorcylate,methyl-2,4-dihydroxybenzoate,unii-xe68bu3oyj,xe68bu3oyj,.beta.-resorcylic acid, methyl ester,beta-resorcylic acid methyl ester,methyl benzoate, 2,4-dihydroxy,methyl b-resorcylate
16523
methyl 2,4-dihydroxybenzoate
COC(=O)C1=C(C=C(C=C1)O)O
Infrared Spectrum Authentic
Melting Point 116°C to 121°C
Quantity 100g
Color White to Beige
Packaging Glass bottle
Assay Percent Range 97%
Linear Formula (HO)2C6H3CO2CH3
Beilstein 10,I,178
Formula Weight 168.15
Physical Form Powder
Percent Purity ≥96.0%
Chemical Name or Material Methyl 2, 4-dihydroxybenzoate
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.