Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,4-dimethoxybenzoate, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
2150-41-6
C10H12O4
196.2
IHIJFZWLGPEYPJ-UHFFFAOYSA-N
2,4-dimethoxybenzoic acid methyl ester,benzoic acid, 2,4-dimethoxy-, methyl ester,acmc-1ccii,methyl2,4-dimethoxybenzoate,methyl 2,4-dimethoxybenzoate,2,4-dimethoxy-benzoic acid methyl ester,benzoic acid,2,4-dimethoxy-,methyl ester,benzoic acid,2,4-dimethoxy-, methyl ester,2,4-dihydroxybenzoic acid, dimethyl ether, methyl ester
137451
methyl 2,4-dimethoxybenzoate
COC1=CC(=C(C=C1)C(=O)OC)OC
2150-41-6
C10H12O4
196.2
IHIJFZWLGPEYPJ-UHFFFAOYSA-N
2,4-dimethoxybenzoic acid methyl ester,benzoic acid, 2,4-dimethoxy-, methyl ester,acmc-1ccii,methyl2,4-dimethoxybenzoate,methyl 2,4-dimethoxybenzoate,2,4-dimethoxy-benzoic acid methyl ester,benzoic acid,2,4-dimethoxy-,methyl ester,benzoic acid,2,4-dimethoxy-, methyl ester,2,4-dihydroxybenzoic acid, dimethyl ether, methyl ester
137451
methyl 2,4-dimethoxybenzoate
COC1=CC(=C(C=C1)C(=O)OC)OC
Flash Point >110°C
Packaging Glass bottle
Specific Gravity 1.194
Density 1.1940g/mL
Boiling Point 294°C to 296°C
Quantity 5g
Assay Percent Range 97.5% min. (GC)
Linear Formula (CH30)2C6H3CO2CH3
Formula Weight 196.2
Percent Purity 98%
Chemical Name or Material Methyl 2, 4-dimethoxybenzoate
Show More Show Less