Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2,5-difluoro-4-nitrobenzoate, 95%

Click to view available options
Quantity:
250mg
1g
5g
924868-81-5
C8H5F2NO4
217.128
MFCD08689670
XBUVRWIIEREYFL-UHFFFAOYSA-N
methyl 2,5-difluoro-4-nitrobenzenecarboxylate,methyl2,5-difluoro-4-nitrobenzoate,benzoic acid, 2,5-difluoro-4-nitro-, methyl ester,methyldifluoronitrobenzenecarboxylate,methyl-2,5-difluoro-4-nitrobenzoate,2,5-difluoro-4-methoxycarbonyl nitrobenzene,2,5-difluoro-4-nitro-benzoic acid methyl ester
18526058
methyl 2,5-difluoro-4-nitrobenzoate
COC(=O)C1=CC(=C(C=C1F)[N+](=O)[O-])F
924868-81-5
C8H5F2NO4
217.128
MFCD08689670
XBUVRWIIEREYFL-UHFFFAOYSA-N
methyl 2,5-difluoro-4-nitrobenzenecarboxylate,methyl2,5-difluoro-4-nitrobenzoate,benzoic acid, 2,5-difluoro-4-nitro-, methyl ester,methyldifluoronitrobenzenecarboxylate,methyl-2,5-difluoro-4-nitrobenzoate,2,5-difluoro-4-methoxycarbonyl nitrobenzene,2,5-difluoro-4-nitro-benzoic acid methyl ester
18526058
methyl 2,5-difluoro-4-nitrobenzoate
COC(=O)C1=CC(=C(C=C1F)[N+](=O)[O-])F
Quantity 1g
Formula Weight 217.13
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Methyl 2,5-difluoro-4-nitrobenzoate

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold