Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2,6-dichlorophenylacetate, 99%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
54551-83-6
C9H8Cl2O2
219.07
MFCD00191640
FCWRUYPZZJPCCG-UHFFFAOYSA-N
methyl 2,6-dichlorophenylacetate,methyl 2-2,6-dichlorophenyl acetate,2,6-dichlorophenylacetic acid methyl ester,benzeneacetic acid,2,6-dichloro-, methyl ester,benzeneacetic acid, 2,6-dichloro-, methyl ester,acmc-209lhf,methyl2,6-dichlorophenylacetate,guanfacine methyl ester impurity,methyl 2,6-dichlorophenyl acetate,2,6-dichloro-phenylacetic acid methyl ester
2734107
methyl 2-(2,6-dichlorophenyl)acetate
COC(=O)CC1=C(C=CC=C1Cl)Cl
54551-83-6
C9H8Cl2O2
219.07
MFCD00191640
FCWRUYPZZJPCCG-UHFFFAOYSA-N
methyl 2,6-dichlorophenylacetate,methyl 2-2,6-dichlorophenyl acetate,2,6-dichlorophenylacetic acid methyl ester,benzeneacetic acid,2,6-dichloro-, methyl ester,benzeneacetic acid, 2,6-dichloro-, methyl ester,acmc-209lhf,methyl2,6-dichlorophenylacetate,guanfacine methyl ester impurity,methyl 2,6-dichlorophenyl acetate,2,6-dichloro-phenylacetic acid methyl ester
2734107
methyl 2-(2,6-dichlorophenyl)acetate
COC(=O)CC1=C(C=CC=C1Cl)Cl
Specific Gravity 1.33
Refractive Index 1.5400 to 1.542
Density 1.3300g/mL
Infrared Spectrum Authentic
Quantity 25g
Color Colorless
Packaging Glass bottle
Assay Percent Range 98.5% min. (GC)
Formula Weight 219.07
Physical Form Liquid
Percent Purity 99%
Chemical Name or Material Methyl 2, 6-dichlorophenylacetate, 99%
Show More Show Less

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning

EINECSNumber : 259-221-8