Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-Acetamidopropionate 98.0+%, TCI America™

Click to view available options
Quantity:
5g
25g
26629-33-4
C6H11NO3
145.158
MFCD01631289
FQGVVDYNRHNTCK-UHFFFAOYSA-N
2-Acetamidopropionic Acid Methyl Ester, N-Acetyl-DL-alanine Methyl Ester
520278
methyl 2-acetamidopropanoate
CC(C(=O)OC)NC(=O)C
26629-33-4
C6H11NO3
145.158
MFCD01631289
FQGVVDYNRHNTCK-UHFFFAOYSA-N
2-Acetamidopropionic Acid Methyl Ester, N-Acetyl-DL-alanine Methyl Ester
520278
methyl 2-acetamidopropanoate
CC(C(=O)OC)NC(=O)C
Quantity 5g
Color White
Melting Point 48°C
Formula Weight 145.16
Physical Form Crystal-Powder at 20°C
Percent Purity ≥98.0% (GC)
Chemical Name or Material Methyl 2-Acetamidopropionate