Learn More
Accela Chembio IncSupplier Diversity Partner METHYL 2-AMINO-4,5-DIFLUO 100G
Manufacturer:  Accela Chembio IncSupplier Diversity Partner SY014552100G
Encompass_Preferred
Methyl 2-amino-4,5-difluorobenzoate, 100g, 207346-42-7, MFCD09264511, 97+%, Shelf Life: 1260 Days, Light Sensitive
Catalog No. 50-252-036