Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-amino-4,5-difluorobenzoate, 98%

Click to view available options
Quantity:
5g
25g
100g
207346-42-7
C8H7F2NO2
187.146
MFCD09264511
VIAQNTRKUPBQKR-UHFFFAOYSA-N
2-amino-4,5-difluorobenzoic acid methyl ester,benzoic acid, 2-amino-4,5-difluoro-, methyl ester,pubchem4676,acmc-209fdn,ksc496g7j,methyl 4,5-difluoroanthranilate,methyl2-amino-4,5-difluorobenzoate,methyl 2-amino-4,5-difluoro-benzoate,2-amino-4,5-difluoro-benzoic acid methyl ester
18472001
methyl 2-amino-4,5-difluorobenzoate
COC(=O)C1=CC(=C(C=C1N)F)F
207346-42-7
C8H7F2NO2
187.146
MFCD09264511
VIAQNTRKUPBQKR-UHFFFAOYSA-N
2-amino-4,5-difluorobenzoic acid methyl ester,benzoic acid, 2-amino-4,5-difluoro-, methyl ester,pubchem4676,acmc-209fdn,ksc496g7j,methyl 4,5-difluoroanthranilate,methyl2-amino-4,5-difluorobenzoate,methyl 2-amino-4,5-difluoro-benzoate,2-amino-4,5-difluoro-benzoic acid methyl ester
18472001
methyl 2-amino-4,5-difluorobenzoate
COC(=O)C1=CC(=C(C=C1N)F)F
Color White
Quantity 25g
Beilstein 2834936
Formula Weight 187.15
Physical Form Solid
Percent Purity 98%
Chemical Name or Material Methyl 2-amino-4,5-difluorobenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning