Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate, 95%, Thermo Scientific™

Catalog Number p-4813542
Click to view available options
Quantity:
1 g
5 g
184174-81-0
C11H15NO2S
225.31
BDPLKEMLVDUYRR-UHFFFAOYSA-N
methyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylate, methyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta-b thiophene-3-carboxylate, methyl 2-amino-4h,5h,6h,7h,8h-cyclohepta b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylic acid methyl ester, 5,6,7,8-tetrahydro-2-amino-4h-cyclohepta b thiophene-3-carboxylic acid methyl ester, methyl 2-amino-4,5,6,7,8-pentahydrocyclohepta 1,2-b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b-thiophene-3-carboxylic acid methyl ester
2756554
methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
COC(=O)C1=C(SC2=C1CCCCC2)N
184174-81-0
C11H15NO2S
225.31
BDPLKEMLVDUYRR-UHFFFAOYSA-N
methyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylate, methyl 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta-b thiophene-3-carboxylate, methyl 2-amino-4h,5h,6h,7h,8h-cyclohepta b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b thiophene-3-carboxylic acid methyl ester, 5,6,7,8-tetrahydro-2-amino-4h-cyclohepta b thiophene-3-carboxylic acid methyl ester, methyl 2-amino-4,5,6,7,8-pentahydrocyclohepta 1,2-b thiophene-3-carboxylate, 2-amino-5,6,7,8-tetrahydro-4h-cyclohepta b-thiophene-3-carboxylic acid methyl ester
2756554
methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
COC(=O)C1=C(SC2=C1CCCCC2)N
Melting Point 63.0°C to 67.0°C
Assay Percent Range 94% min. (HPLC)
Packaging Glass Bottle
Quantity 1 g
Formula Weight 225.31
Percent Purity 95%
Chemical Name or Material Methyl 2-amino-5,6,7,8-tetrahydro-4H-cyclohepta[b]thiophene-3-carboxylate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit