Alfa Aesar™ Methyl 2-benzyloxybenzoate, 98%

Manufacturer: Alfa Aesar™ B2330922

Quantity 100g

 View more versions of this product

Catalog No. AAB2330922

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 55142-16-0
    Molecular Formula C15H14O3
    Formula Weight 242.27
    MDL Number MFCD01099045
    Synonym 2-Benzyloxybenzoic acid methyl ester
    Chemical Name or Material Methyl 2-benzyloxybenzoate

    Specifications

    Formula Weight 242.27
    Molecular Formula C15H14O3
    MDL Number MFCD01099045
    Synonym 2-Benzyloxybenzoic acid methyl ester
    Melting Point 45° to 48°C
    Quantity 100g
    CAS 55142-16-0
    Chemical Name or Material Methyl 2-benzyloxybenzoate
    Assay 98%

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit