Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-benzyloxybenzoate, 98%

Click to view available options
Quantity:
5g
25g
100g
55142-16-0
C15H14O3
242.274
MFCD01099045
FULHVBYAYSGTKX-UHFFFAOYSA-N
methyl 2-benzyloxybenzoate,methyl 2-benzyloxy benzoate,2-benzyloxybenzoic acid methyl ester,methyl 2-phenylmethoxy benzoate,methyl 2-benzyloxy-benzoate,acmc-1b03z,methyl salicylate monobenzyl ether,2-benzyloxy benzoic acid methyl ester,2-phenylmethoxybenzoic acid methyl ester,benzoic acid,2-phenylmethoxy-, methyl ester
251331
methyl 2-phenylmethoxybenzoate
COC(=O)C1=CC=CC=C1OCC2=CC=CC=C2
55142-16-0
C15H14O3
242.274
MFCD01099045
FULHVBYAYSGTKX-UHFFFAOYSA-N
methyl 2-benzyloxybenzoate,methyl 2-benzyloxy benzoate,2-benzyloxybenzoic acid methyl ester,methyl 2-phenylmethoxy benzoate,methyl 2-benzyloxy-benzoate,acmc-1b03z,methyl salicylate monobenzyl ether,2-benzyloxy benzoic acid methyl ester,2-phenylmethoxybenzoic acid methyl ester,benzoic acid,2-phenylmethoxy-, methyl ester
251331
methyl 2-phenylmethoxybenzoate
COC(=O)C1=CC=CC=C1OCC2=CC=CC=C2
Quantity 25g
Melting Point 45°C to 48°C
Formula Weight 242.27
Percent Purity 98%
Chemical Name or Material Methyl 2-benzyloxybenzoate