Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, 95%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
88568-95-0
C13H18NO7P
331.261
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate,+/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester,n-cbz-2-phosphonoglycine trimethyl ester,n-benzyloxy carbonyl,+/--cbz-alpha-phosphonoglycine trimethyl ester,methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate,+/--n-cbz-alpha-phosphonoglycine trimethyl ester,z-alpha-phosphonoglycine trimethyl ester,cbz-alpha-phosphono-dl-glycine trimethyl ester
2734718
methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
88568-95-0
C13H18NO7P
331.261
MFCD00043304
GSYSFVSGPABNNL-UHFFFAOYSA-N
methyl 2-benzyloxy carbonyl amino-2-dimethoxyphosphoryl acetate,+/--benzyloxycarbonyl-alpha-phosphonoglycine trimethyl ester,n-cbz-2-phosphonoglycine trimethyl ester,n-benzyloxy carbonyl,+/--cbz-alpha-phosphonoglycine trimethyl ester,methyl 2-benzyloxycarbonylamino-2-dimethoxyphosphoryl-acetate,+/--n-cbz-alpha-phosphonoglycine trimethyl ester,z-alpha-phosphonoglycine trimethyl ester,cbz-alpha-phosphono-dl-glycine trimethyl ester
2734718
methyl 2-dimethoxyphosphoryl-2-(phenylmethoxycarbonylamino)acetate
COC(=O)C(NC(=O)OCC1=CC=CC=C1)P(=O)(OC)OC
for C-C Bond Formation (Olefination)
Color White
Infrared Spectrum Authentic
Quantity 1g
Melting Point 77°C to 80°C
CAS Min % 94.0
CAS Max % 100.0
Assay Percent Range 95%
Formula Weight 331.26
Physical Form Crystalline Powder
Percent Purity ≥94%
Chemical Name or Material ±-Methyl 2-benzyloxycarbonylamino-2-(dimethoxyphosphinyl)acetate, 95%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.