Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-bromo-5-chlorobenzoate, 98%

Click to view available options
Quantity:
25g
100g
27007-53-0
C8H6BrClO2
249.488
MFCD00144763
BIECSXCXIXHDBC-UHFFFAOYSA-N
2-bromo-5-chlorobenzoic acid methyl ester,benzoic acid, 2-bromo-5-chloro-, methyl ester,pubchem11238,acmc-1cr8r,ksc494q3f,methyl-2-bromo-5-chlorobenzoate,methyl 2-bromo-5-chloro-benzoate,methyl 2-bromanyl-5-chloranyl-benzoate,2-bomo-5-chlorobenzoic acid methyl ester
280500
methyl 2-bromo-5-chlorobenzoate
COC(=O)C1=C(C=CC(=C1)Cl)Br
27007-53-0
C8H6BrClO2
249.488
MFCD00144763
BIECSXCXIXHDBC-UHFFFAOYSA-N
2-bromo-5-chlorobenzoic acid methyl ester,benzoic acid, 2-bromo-5-chloro-, methyl ester,pubchem11238,acmc-1cr8r,ksc494q3f,methyl-2-bromo-5-chlorobenzoate,methyl 2-bromo-5-chloro-benzoate,methyl 2-bromanyl-5-chloranyl-benzoate,2-bomo-5-chlorobenzoic acid methyl ester
280500
methyl 2-bromo-5-chlorobenzoate
COC(=O)C1=C(C=CC(=C1)Cl)Br
Applications
Methyl 2-bromo-5-chlorobenzoate is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
Quantity 25g
Melting Point 37°C to 40°C
Formula Weight 249.49
Percent Purity 98%
Chemical Name or Material Methyl 2-bromo-5-chlorobenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning