Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2-bromomethyl-3-nitrobenzoate, 95%

Click to view available options
Quantity:
250mg
1g
98475-07-1
C9H8BrNO4
274.07
MFCD04114315
FCGIVHSBEKGQMZ-UHFFFAOYSA-N
methyl 2-bromomethyl-3-nitrobenzoate,2-bromomethyl-3-nitrobenzoic acid methyl ester,methyl 2-bromomethyl-3-nitro-benzoate,2-bromomethyl-3-nitrobenzoic acid methyl ester,benzoic acid, 2-bromomethyl-3-nitro-, methyl ester,methyl 2-bromomethyl-3-nitrobenzenecarboxylate,2-bromomethyl-3-nitro benzoic acid methyl ester,methyl2-bromomethyl-3-nitrobenzoate,zlchem 474,pubchem19169
1487795
methyl 2-(bromomethyl)-3-nitrobenzoate
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])CBr
98475-07-1
C9H8BrNO4
274.07
MFCD04114315
FCGIVHSBEKGQMZ-UHFFFAOYSA-N
methyl 2-bromomethyl-3-nitrobenzoate,2-bromomethyl-3-nitrobenzoic acid methyl ester,methyl 2-bromomethyl-3-nitro-benzoate,2-bromomethyl-3-nitrobenzoic acid methyl ester,benzoic acid, 2-bromomethyl-3-nitro-, methyl ester,methyl 2-bromomethyl-3-nitrobenzenecarboxylate,2-bromomethyl-3-nitro benzoic acid methyl ester,methyl2-bromomethyl-3-nitrobenzoate,zlchem 474,pubchem19169
1487795
methyl 2-(bromomethyl)-3-nitrobenzoate
COC(=O)C1=C(C(=CC=C1)[N+](=O)[O-])CBr
Quantity 1g
Beilstein 7811923
Formula Weight 274.07
Physical Form Solid
Percent Purity 95%
Chemical Name or Material Methyl 2-bromomethyl-3-nitrobenzoate