Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-(bromomethyl)acrylate, 96%, Acros Organics™

Click to view available options
Quantity:
1g
5g
25g
4224-69-5
C5H7BrO2
179.01
CFTUQSLVERGMHL-UHFFFAOYSA-N
methyl 2-bromomethyl acrylate,methyl 2-bromomethyl prop-2-enoate,methyl 2-bromomethyl-2-propenoate,methyl alpha-bromomethyl acrylate,2-bromomethyl methyl acrylate,2-propenoic acid, 2-bromomethyl-, methyl ester,2-bromomethyl acrylic acid methyl ester,acrylic acid, 2-bromomethyl-, methyl ester,zlchem 195,pubchem17371
521093
methyl 2-(bromomethyl)prop-2-enoate
COC(=O)C(=C)CBr
4224-69-5
C5H7BrO2
179.01
CFTUQSLVERGMHL-UHFFFAOYSA-N
methyl 2-bromomethyl acrylate,methyl 2-bromomethyl prop-2-enoate,methyl 2-bromomethyl-2-propenoate,methyl alpha-bromomethyl acrylate,2-bromomethyl methyl acrylate,2-propenoic acid, 2-bromomethyl-, methyl ester,2-bromomethyl acrylic acid methyl ester,acrylic acid, 2-bromomethyl-, methyl ester,zlchem 195,pubchem17371
521093
methyl 2-(bromomethyl)prop-2-enoate
COC(=O)C(=C)CBr
Quantity 1g
Packaging Glass bottle
Boiling Point 35.0°C to 37.0°C (13.0mmHg)
Density 1.4890g/mL
Color Brown to Colorless
Specific Gravity 1.489
Flash Point 78°C
Infrared Spectrum Authentic
Refractive Index 1.4870 to 1.4890
Assay Percent Range 95% min. (GC)
Formula Weight 179.01
Physical Form Liquid
Percent Purity 96%
Chemical Name or Material Methyl 2-(bromomethyl)acrylate
Show More Show Less

GHS H Statement
Causes serious eye damage.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.

GHS Signal Word: Danger